Assay Details
Target Gene Details
Entrez Gene ID: | 50927 |
Gene Name: | nuclear autoantigenic sperm protein (histone-binding) |
Gene Aliases: |
5033430J04Rik, AI131596, AI317140, D4Ertd767e, Epcs32, Nasp-T |
Location: |
Chr.4:116601052-116627951 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nasp | NM_001081475.1 | 14 | 1873 | NP_001074944.1 |
| NM_001284229.1 | 13 | 1792 | NP_001271158.1 | |
| NM_016777.3 | 16 | 2842 | NP_058057.3 | |
| AK083333.1 | 14 | 1874 | BAC38871.1 | |
| BM243364.2 | 1 | 246 |
More Information
Set Membership: |
|

Genomic Map