Assay Details
Target Gene Details
Entrez Gene ID: | 12395 |
Gene Name: | runt-related transcription factor 1; translocated to, 1 (cyclin D-related) |
Gene Aliases: |
Cbfa2t1h, ETO, MTG8 |
Location: |
Chr.4:13743182-13895056 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Runx1t1 | NM_001111026.2 | 11 | 4571 | NP_001104496.1 |
| NM_001111027.2 | 12 | 4765 | NP_001104497.1 | |
| NM_009822.3 | 11 | 4503 | NP_033952.1 | |
| AK134476.1 | 1 | 1458 | BAE22156.1 | |
| AK162711.1 | 1 | 125 |
More Information
Set Membership: |
|

Genomic Map