Assay Details
Target Gene Details
Entrez Gene ID: | 83771 |
Gene Name: | taste receptor, type 1, member 3 |
Gene Aliases: |
Sac, T1r3 |
Location: |
Chr.4:155859268-155863353 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas1r3 | NM_031872.2 | 3 | 1050 | NP_114078.1 |
| AB049994.1 | 3 | 1063 | BAB47181.1 | |
| AF311386.1 | 3 | 1031 | AAL08425.1 | |
| AF337039.1 | 3 | 1050 | AAK39436.1 | |
| AY032621.1 | 3 | 1031 | AAK51602.1 |
More Information
Set Membership: |
|

Genomic Map