Assay Details
Target Gene Details
Entrez Gene ID: | 22384 |
Gene Name: | eukaryotic translation initiation factor 4H |
Gene Aliases: |
AU018978, D5Ertd355e, E430026L18Rik, Ef4h, Wbscr1, Wscr1, mKIAA0038 |
Location: |
Chr.5:134619876-134639494 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4h | NM_033561.1 | 7 | 1519 | NP_291039.1 |
| AK045004.1 | 7 | 1521 | BAC32180.1 | |
| AK088818.1 | 7 | 1531 | BAC40593.1 | |
| AK122194.1 | 5 | 4741 | BAC65476.3 | |
| AK132034.1 | 7 | 1539 | BAE20955.1 | |
| AK135390.1 | 7 | 1520 | BAE22517.1 | |
| AK136999.1 | 6 | 1446 | BAE23201.1 | |
| AK149841.1 | 7 | 1505 | BAE29116.1 | |
| AK150088.1 | 7 | 1528 | BAE29297.1 | |
| AK166567.1 | 7 | 1520 | BAE38858.1 | |
| AK168872.1 | 6 | 1478 | BAE40690.1 | |
| AK169312.1 | 7 | 1502 | BAE41067.1 | |
| AK169353.1 | 7 | 1520 | BAE41102.1 | |
| AK169407.1 | 6 | 1478 | BAE41153.1 | |
| AK191915.1 | ||||
| BC014796.1 | 6 | 1473 | AAH14796.1 |
More Information
Set Membership: |
|

Genomic Map