Assay Details
Target Gene Details
Entrez Gene ID: | 12854 |
Gene Name: | cortistatin |
Gene Aliases: |
CST, PCST |
Location: |
Chr.4:149125191-149126741 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cort | NM_007745.3 | 2 | 358 | NP_031771.1 |
| AF013253.1 | 2 | 279 | AAB66896.1 | |
| AK013496.1 | 2 | 360 | BAB28882.1 |
More Information
Set Membership: |
|

Genomic Map