Assay Details
Target Gene Details
Entrez Gene ID: | 56031 |
Gene Name: | peptidylprolyl isomerase E (cyclophilin E) |
Gene Aliases: |
2010010D16Rik, Cyp33 |
Location: |
Chr.4:123127114-123140000 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppie | NM_019489.5 | 10 | 1113 | NP_062362.1 |
| AK008177.1 | 10 | 1114 | BAB25512.1 | |
| AK042165.1 | 7 | 3697 | ||
| BC045154.1 | 10 | 1108 | AAH45154.1 |
More Information
Set Membership: |
|

Genomic Map