Assay Details
Target Gene Details
Entrez Gene ID: | 14673 |
Gene Name: | guanine nucleotide binding protein, alpha 12 |
Gene Aliases: |
AI414047, AI504261, Galpha12 |
Location: |
Chr.5:140759944-140830431 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna12 | NM_010302.2 | 4 | 1490 | NP_034432.1 |
| M63659.1 | 4 | 1490 | AAA37648.1 |
More Information
Set Membership: |
|

Genomic Map