Assay Details
Target Gene Details
Entrez Gene ID: | 21899 |
Gene Name: | toll-like receptor 6 |
Gene Aliases: |
- |
Location: |
Chr.5:64952030-64971173 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlr6 | NM_011604.3 | 2 | 516 | NP_035734.3 |
| AB020808.1 | 1 | 404 | BAA78632.1 | |
| AF314636.1 | 1 | 384 | AAG38563.1 | |
| AK137502.1 | 2 | 471 | BAE23384.1 | |
| AK154253.1 | 2 | 442 | ||
| BC055366.1 | 2 | 529 | AAH55366.1 | |
| DQ790887.1 | 1 | 384 |

Genomic Map