Assay Details
Target Gene Details
Entrez Gene ID: | 258569 |
Gene Name: | olfactory receptor 459 |
Gene Aliases: |
MOR120-1 |
Location: |
Chr.6:41771353-41772297 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr459 | NM_146576.1 | 1 | 635 | NP_666787.1 |
| BC116766.1 | 1 | 704 | ||
| BC120604.1 | 1 | 704 |
More Information
Set Membership: |
|

Genomic Map