Assay Details
Target Gene Details
Entrez Gene ID: | 60321 |
Gene Name: | WW domain binding protein 11 |
Gene Aliases: |
2510026P17Rik, D6Wsu113e, Npwbp, SIPP1 |
Location: |
Chr.6:136813654-136828216 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbp11 | NM_021714.4 | 10 | 1405 | NP_068360.4 |
AF071186.1 | 4 | 531 | AAC34812.1 | |
AK031678.1 | 10 | 1230 | BAC27508.1 | |
AK041079.1 | 10 | 1344 | BAC30812.1 | |
AK144968.1 | 10 | 1329 | BAE26160.1 | |
AK145917.1 | 10 | 1355 | BAE26748.1 | |
AK151989.1 | 10 | 1351 | BAE30854.1 | |
AK153044.1 | 10 | 1351 | BAE31673.1 | |
AK159257.1 | 10 | 1343 | BAE34938.1 | |
BC006600.1 | 10 | 1317 | AAH06600.1 | |
BC021823.1 | 10 | 1298 | AAH21823.1 | |
BC104145.2 | 1 | 109 |
