Assay Details
Target Gene Details
Entrez Gene ID: | 53324 |
Gene Name: | neuronal pentraxin 2 |
Gene Aliases: |
Narp, np2 |
Location: |
Chr.5:144545887-144557478 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nptx2 | NM_016789.3 | 1 | 196 | NP_058069.1 |
AF049124.1 | 1 | 87 | AAC05131.1 | |
AK134806.1 | 1 | 197 | BAE22293.1 | |
BC026054.1 | 1 | 191 | AAH26054.1 |
