Assay Details
Target Gene Details
Entrez Gene ID: | 18575 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
- |
Location: |
Chr.6:56069798-56652516 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde1c | NM_001159953.1 | 19 | 2881 | NP_001153425.1 |
| AK030423.1 | 19 | 2883 | BAC26956.1 | |
| AK133243.1 | 21 | 2487 |
More Information
Set Membership: |
|

Genomic Map