Assay Details
Target Gene Details
Entrez Gene ID: | 56361 |
Gene Name: | pseudouridine synthase 1 |
Gene Aliases: |
A730013B20Rik, MPUS1, mPus1p |
Location: |
Chr.5:110773667-110781056 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pus1 | NM_001025561.3 | 7 | 1657 | NP_001020732.1 |
| NM_001025562.2 | 6 | 1300 | NP_001020733.1 | |
| NM_019700.4 | 6 | 1603 | NP_062674.2 | |
| AB041563.1 | 7 | 1659 | BAA95047.1 | |
| AF116237.1 | 5 | 1260 | AAD21041.1 | |
| AK089258.1 | 6 | 1603 | BAC40817.1 | |
| AK138570.1 | 4 | 1178 | BAE23701.1 | |
| AK161521.1 | 6 | 1312 | BAE36440.1 | |
| BC021446.1 | 6 | 1479 | ||
| BC034359.1 | 6 | 1574 | ||
| CF618003.1 | 1 | 262 |
More Information
Set Membership: |
|

Genomic Map