Assay Details
Target Gene Details
Entrez Gene ID: | 24116 |
Gene Name: | negative elongation factor complex member A, Whsc2 |
Gene Aliases: |
Whsc2, Whsc2h |
Location: |
Chr.5:33898179-33936258 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfa | NM_011914.2 | 9 | 1108 | NP_036044.1 |
| AF101435.1 | 9 | 1066 | AAC72983.1 | |
| AK019191.1 | 9 | 1174 | ||
| AK078760.1 | 9 | 1108 | BAC37379.1 | |
| AK088384.1 | 9 | 1124 | BAC40319.1 | |
| AK146185.1 | 9 | 1160 | BAE26961.1 | |
| BC038003.1 | 9 | 1109 | AAH38003.1 |

Genomic Map