Assay Details
Target Gene Details
Entrez Gene ID: | 21937 |
Gene Name: | tumor necrosis factor receptor superfamily, member 1a |
Gene Aliases: |
CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI, TNFRp55, TNFalpha-R1, Tnfr-2, Tnfr1, p55, p55-R |
Location: |
Chr.6:125349723-125362483 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf1a | NM_011609.4 | 1 | 113 | NP_035739.2 |
| AK154388.1 | 1 | 83 | BAE32554.1 | |
| AK159989.1 | 1 | 37 | BAE35539.1 | |
| AK181531.1 | 1 | 93 | ||
| BC004599.1 | AAH04599.1 | |||
| BC052675.1 | 1 | 48 | AAH52675.1 | |
| BC096429.1 | 1 | 65 | AAH96429.1 | |
| CK624043.1 | 1 | 113 | ||
| L26349.1 | 1 | 81 | AAA59361.1 | |
| M60468.1 | AAA39751.1 | |||
| X57796.1 | 1 | 84 | CAA40936.1 | |
| X59238.1 | CAA41922.1 |
More Information
Set Membership: |
|

Genomic Map