Assay Details
Target Gene Details
Entrez Gene ID: | 269717 |
Gene Name: | ORAI calcium release-activated calcium modulator 2 |
Gene Aliases: |
A730041O15Rik, Tmem142b |
Location: |
Chr.5:136147459-136170656 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orai2 | NM_178751.3 | 3 | 3265 | NP_848866.2 |
| AK042940.1 | 3 | 3266 | BAC31410.1 | |
| AK190197.1 | 1 | 49 | ||
| BC066070.1 | 3 | 3212 | AAH66070.1 |
More Information
Set Membership: |
|

Genomic Map