Assay Details
Target Gene Details
Entrez Gene ID: | 232016 |
Gene Name: | coiled-coil domain containing 129 |
Gene Aliases: |
D530004J12Rik, Gm458 |
Location: |
Chr.6:55804628-55978743 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc129 | NM_001081665.1 | 11 | 1615 | NP_001075134.1 |
| AK085190.1 | 7 | 1259 | BAC39386.1 | |
| BC116335.2 | 11 | 1615 |

Genomic Map