Assay Details
Target Gene Details
Entrez Gene ID: | 13448 |
Gene Name: | docking protein 1 |
Gene Aliases: |
AW557123, p62DOK |
Location: |
Chr.6:83030932-83033471 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dok1 | NM_001291799.1 | 2 | 249 | NP_001278728.1 |
NM_010070.4 | 2 | 251 | NP_034200.4 | |
AK136382.1 | 2 | 252 | BAE22956.1 | |
AK169871.1 | 2 | 247 | ||
BC013066.1 | 2 | 245 | AAH13066.1 | |
BY114582.1 | 2 | 209 | ||
CJ053718.1 | 2 | 252 | ||
U78818.2 | 2 | 216 | AAB48827.2 |
