Assay Details
Target Gene Details
Entrez Gene ID: | 194352 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 5 |
Gene Aliases: |
CAT2, D630033B11, ECAC1 |
Location: |
Chr.6:41651661-41682393 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv5 | NM_001007572.2 | 13 | 1614 | NP_001007573.1 |
| AK085479.1 | 13 | 1660 | ||
| BC110554.1 | 13 | 1614 | AAI10555.1 | |
| BC110555.1 | 13 | 1614 | AAI10556.1 |
More Information
Set Membership: |
|

Genomic Map