Assay Details
Target Gene Details
Entrez Gene ID: | 672284 |
Gene Name: | NK1 transcription factor related, locus 1 (Drosophila) |
Gene Aliases: |
Nkx-1.1, Sax2 |
Location: |
Chr.5:33430734-33434090 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx1-1 | NM_011320.1 | 1 | 413 | NP_035450.1 |
More Information
Set Membership: |
|

Genomic Map