Assay Details
Target Gene Details
Entrez Gene ID: | 269831 |
Gene Name: | tetraspanin 12 |
Gene Aliases: |
9030619E17, AI426782, AI663988, AW111457, Tm4sf12 |
Location: |
Chr.6:21771391-21853349 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan12 |

Genomic Map