Assay Details
Target Gene Details
Entrez Gene ID: | 330260 |
Gene Name: | paraoxonase 2 |
Gene Aliases: |
6330405I24Rik, AI481612 |
Location: |
Chr.6:5264620-5298373 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pon2 | NM_183308.2 | 9 | 1140 | NP_899131.1 |
| AK007350.1 | 9 | 1109 | ||
| AK146311.1 | 9 | 1108 | BAE27066.1 | |
| AK152351.1 | 9 | 1091 | ||
| AK171926.1 | 9 | 1098 | BAE42735.1 | |
| AK181604.1 | 1 | 140 | ||
| AK188334.1 | 1 | 140 | ||
| AK211087.1 | 1 | 140 | ||
| AK211348.1 | 1 | 49 | ||
| AK211680.1 | 1 | 67 | ||
| AK212729.1 | ||||
| AK213848.1 | 1 | 140 | ||
| AK219612.1 | 1 | 140 | ||
| BC037140.1 | 9 | 1082 | AAH37140.1 | |
| BC055896.1 | 9 | 1140 | AAH55896.1 | |
| BC062200.1 | 9 | 1131 | AAH62200.1 | |
| L48514.1 | 9 | 1076 | AAC42089.1 |

Genomic Map