Assay Details
Target Gene Details
Entrez Gene ID: | 68089 |
Gene Name: | actin related protein 2/3 complex, subunit 4 |
Gene Aliases: |
20kDa, 5330419I20Rik, AI327076, p20-Arc |
Location: |
Chr.6:113378113-113390447 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc4 | NM_001170485.1 | 4 | 986 | NP_001163956.1 |
| NM_001170486.1 | 5 | 1098 | NP_001163957.1 | |
| NM_026552.3 | 6 | 1217 | NP_080828.1 | |
| AJ278129.1 | 6 | 1143 | CAC34583.1 | |
| AK003923.1 | 1 | 300 | ||
| AK017993.1 | 6 | 1218 | BAB31026.1 | |
| AK018069.1 | 6 | 1155 | BAB31059.1 | |
| AK039215.1 | 6 | 1165 | BAC30279.1 | |
| AK076248.1 | 6 | 1165 | BAC36272.1 | |
| AK077325.1 | 6 | 1139 | BAC36751.1 | |
| AK133626.1 | 6 | 1165 | BAE21755.1 | |
| AK136082.1 | 6 | 1160 | BAE22810.1 | |
| AK150690.1 | 6 | 1151 | BAE29769.1 | |
| AK153380.1 | 6 | 1149 | BAE31946.1 | |
| AK159410.1 | 5 | 1046 | BAE35061.1 | |
| AK166440.1 | 6 | 1139 | BAE38777.1 | |
| BC055309.1 | 5 | 1128 | AAH55309.1 |
Target Gene Details
Entrez Gene ID: | 101100 |
Gene Name: | tubulin tyrosine ligase-like family, member 3 |
Gene Aliases: |
4833441J24Rik, AI450050 |
Location: |
Chr.6:113389260-113414583 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll3 | NM_001142732.1 | 1 | 134 | NP_001136204.1 |
| AK029462.1 | 1 | 135 | BAC26459.1 |
More Information
Set Membership: |
|

Genomic Map