Assay Details
Target Gene Details
Entrez Gene ID: | 17193 |
Gene Name: | methyl-CpG binding domain protein 4 |
Gene Aliases: |
Med1 |
Location: |
Chr.6:115840697-115853438 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbd4 | NM_010774.2 | 3 | 572 | NP_034904.2 |
| AF072249.1 | 3 | 589 | AAC68878.1 | |
| AK148171.1 | 3 | 520 | BAE28391.1 | |
| AK161005.1 | 3 | 1048 | BAE36144.1 | |
| BC024812.1 | 3 | 554 | AAH24812.1 | |
| BQ885775.1 | 3 | 572 |
More Information
Set Membership: |
|

Genomic Map