Assay Details
Target Gene Details
Entrez Gene ID: | 17193 |
Gene Name: | methyl-CpG binding domain protein 4 |
Gene Aliases: |
Med1 |
Location: |
Chr.6:115840697-115853438 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbd4 | NM_010774.2 | 8 | 2038 | NP_034904.2 |
| AK147782.1 | 5 | 1020 | BAE28134.1 |
More Information
Set Membership: |
|

Genomic Map