Assay Details
Target Gene Details
Entrez Gene ID: | 17313 |
Gene Name: | matrix Gla protein |
Gene Aliases: |
Mglap |
Location: |
Chr.6:136872435-136875805 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgp | NM_008597.3 | 4 | 303 | NP_032623.1 |
| AK018719.1 | 4 | 304 | BAB31365.1 | |
| AK159591.1 | 4 | 307 | BAE35211.1 | |
| AK159931.1 | 4 | 304 | BAE35492.1 | |
| BC018338.1 | 4 | 195 | AAH18338.1 | |
| BC036991.1 | 4 | 299 | AAH36991.1 | |
| BC069922.1 | 4 | 293 | AAH69922.1 | |
| BC079478.1 | 4 | 294 | AAH79478.1 | |
| BY702829.1 | 4 | 304 | ||
| D00613.1 | 4 | 246 | BAA00488.1 |

Genomic Map