Assay Details
Target Gene Details
Entrez Gene ID: | 16511 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 2 |
Gene Aliases: |
AI326795, ERG1, LQT, Lqt2, M-erg, Merg1, merg1a, merg1b |
Location: |
Chr.5:24319589-24351604 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh2 | NM_013569.2 | 4 | 1019 | NP_038597.2 |
| AF012868.1 | 4 | 1025 | AAC53418.1 | |
| AY374424.1 | 4 | 704 | AAQ82708.1 | |
| BC051016.1 | 4 | 850 | AAH51016.1 |
More Information
Set Membership: |
|

Genomic Map