Assay Details
Target Gene Details
Entrez Gene ID: | 231051 |
Gene Name: | lysine (K)-specific methyltransferase 2C |
Gene Aliases: |
E330008K23Rik, HALR, Mll3, mKIAA1506 |
Location: |
Chr.5:25271794-25498818 on Build GRCm38 |
Assay Gene Location: | Within Exon 59 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kmt2c | NM_001081383.1 | 59 | 15839 | NP_001074852.1 |
AK077567.1 | 4 | 1198 | BAC36867.1 | |
AK129377.1 | 18 | 5454 | BAC98187.1 | |
AK148595.1 | 8 | 1847 | ||
AK161051.1 | 8 | 2584 | BAE36172.1 | |
AK164521.1 | 8 | 2103 | BAE37820.1 |
