Assay Details
Target Gene Details
Entrez Gene ID: | 18609 |
Gene Name: | pancreatic and duodenal homeobox 1 |
Gene Aliases: |
IDX-1, IPF-1, Ipf1, Mody4, STF-1, pdx-1 |
Location: |
Chr.5:147270131-147275149 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdx1 | NM_008814.3 | 2 | 1154 | NP_032840.1 |
BC103572.1 | 2 | 1154 | AAI03573.1 | |
BC103581.1 | 2 | 1154 | AAI03582.1 | |
BC103582.1 | 2 | 1154 | AAI03583.1 | |
BC105642.1 | 2 | 1154 | ||
X74342.1 | 2 | 1325 | CAA52389.1 |
