Assay Details
Target Gene Details
Entrez Gene ID: | 57377 |
Gene Name: | mannosyl-oligosaccharide glucosidase |
Gene Aliases: |
1810017N02Rik, AI181835, Gcs1 |
Location: |
Chr.6:83115506-83118902 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mogs | NM_020619.2 | 4 | 2126 | NP_065644.2 |
| AK007528.1 | 1 | 330 | ||
| AK144946.1 | 4 | 2127 | BAE26149.1 | |
| AK150873.1 | 1 | 862 | ||
| AK151941.1 | 1 | 862 | BAE30817.1 | |
| AK152127.1 | 1 | 862 | BAE30968.1 | |
| AK157286.1 | 4 | 2123 | BAE34029.1 | |
| BC051949.1 | 4 | 2126 | AAH51949.1 |
More Information
Set Membership: |
|

Genomic Map