Assay Details
Target Gene Details
Entrez Gene ID: | 20901 |
Gene Name: | serine/threonine kinase receptor associated protein |
Gene Aliases: |
AW557906, C78091, C79202, Unrip |
Location: |
Chr.6:137735082-137751930 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strap | NM_011499.3 | 10 | 1493 | NP_035629.2 |
| AF096285.1 | 11 | 1308 | AAC98300.1 | |
| AK075804.1 | 10 | 1451 | BAC35972.1 | |
| AK077504.1 | 10 | 1445 | BAC36834.1 | |
| AK145971.1 | 10 | 1445 | BAE26796.1 | |
| AK147179.1 | 10 | 1495 | ||
| AK152283.1 | 10 | 1343 | BAE31097.1 | |
| AK154158.1 | 10 | 1462 | ||
| AK199765.1 | 1 | 106 | ||
| BC141076.1 | 10 | 1444 | ||
| BC145342.1 | 10 | 1444 |
More Information
Set Membership: |
|

Genomic Map