Assay Details
Target Gene Details
Entrez Gene ID: | 16511 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 2 |
Gene Aliases: |
AI326795, ERG1, LQT, Lqt2, M-erg, Merg1, merg1a, merg1b |
Location: |
Chr.5:24319589-24351604 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh2 | NM_001294162.1 | 3 | 990 | NP_001281091.1 |
| NM_013569.2 | 7 | 2018 | NP_038597.2 | |
| AF012868.1 | 7 | 2024 | AAC53418.1 | |
| AF012869.1 | 3 | 938 | AAC53419.1 | |
| AF034762.1 | 3 | 982 | AAB87571.1 | |
| AY374424.1 | 7 | 1703 | AAQ82708.1 | |
| BC051016.1 | 7 | 1849 | AAH51016.1 | |
| BM936394.1 | 2 | 355 |
More Information
Set Membership: |
|

Genomic Map