Assay Details
Target Gene Details
Entrez Gene ID: | 77574 |
Gene Name: | TRPM8 channel-associated factor 1 |
Gene Aliases: |
2810407D09Rik, 3321401G04Rik, A230020K05Rik, AI848529, Fam115a |
Location: |
Chr.6:42667999-42710110 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcaf1 | NM_029930.2 | 3 | 1585 | NP_084206.1 |
AK014379.1 | 3 | 1115 | BAC25433.1 | |
AK053509.1 | 3 | 1586 | BAC35410.1 | |
AK076242.1 | 3 | 1531 | ||
AK083917.1 | 3 | 1533 | BAC39061.1 | |
AK132148.1 | 3 | 1553 | BAE20997.1 | |
AK148593.1 | 3 | 1564 | ||
AK173013.1 | 3 | 1567 | BAD32291.1 | |
BC031718.1 | 3 | 1613 | AAH31718.1 |
