Assay Details
Target Gene Details
Entrez Gene ID: | 56284 |
Gene Name: | mitochondrial ribosomal protein L19 |
Gene Aliases: |
9030416F12Rik, 9130412E02Rik, D6Ertd157e, MRP-L15, RLX1, Rpml15 |
Location: |
Chr.6:81957826-81965949 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl19 | NM_026490.2 | 5 | 2848 | NP_080766.1 |
| AK030793.1 | 5 | 2940 |
More Information
Set Membership: |
|

Genomic Map