Assay Details
Target Gene Details
Entrez Gene ID: | 14548 |
Gene Name: | mitochondrial ribosomal protein S33 |
Gene Aliases: |
AI841153, CGI-139, Gdap3, MRP-S33, PTD003 |
Location: |
Chr.6:39801804-39810936 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps33 | NM_001010930.1 | 3 | 574 | NP_001010930.1 |
| NM_010270.2 | 3 | 690 | NP_034400.1 | |
| AI841153.1 | ||||
| AK019000.1 | BAB31503.1 | |||
| AK144131.1 | 3 | 647 | BAE25718.1 | |
| BC028645.1 | AAH28645.1 | |||
| BC092135.1 | AAH92135.1 | |||
| BC096397.1 | AAH96397.1 | |||
| Y17852.1 | CAA76895.1 |
More Information
Set Membership: |
|

Genomic Map