Assay Details
Target Gene Details
Entrez Gene ID: | 19242 |
Gene Name: | pleiotrophin |
Gene Aliases: |
HARP, HB-GAM, HBBN, HBGF-8, HBNF, OSF, Osf-1, Osf1 |
Location: |
Chr.6:36714909-36811361 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptn | NM_008973.2 | 1 | 256 | NP_032999.1 |
More Information
Set Membership: |
|

Genomic Map