Assay Details
Target Gene Details
Entrez Gene ID: | 67861 |
Gene Name: | aldo-keto reductase family 1, member B10 (aldose reductase) |
Gene Aliases: |
2310005E10Rik, Akr1b16 |
Location: |
Chr.6:34384206-34396954 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b10 | NM_172398.3 | 10 | 1058 | NP_765986.3 |
| AK019906.1 | 10 | 954 | ||
| AK075865.1 | 10 | 1060 | ||
| AK082777.1 | 10 | 1016 | BAC38615.1 | |
| AK087002.1 | 10 | 1044 | ||
| BC037690.1 | 10 | 1065 | AAH37690.1 | |
| BY430637.1 | 2 | 105 |
More Information
Set Membership: |
|

Genomic Map