Assay Details
Target Gene Details
Entrez Gene ID: | 108151 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D |
Gene Aliases: |
4631426B19Rik |
Location: |
Chr.5:12383166-12588943 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3d | NM_028882.4 | 18 | 4269 | NP_083158.3 |
| AK014535.1 | 1 | 1533 | ||
| AK031704.1 | 18 | 3881 | BAC27522.1 | |
| AK163828.1 | 3 | 2061 | BAE37508.1 |
More Information
Set Membership: |
|

Genomic Map