Assay Details
Target Gene Details
Entrez Gene ID: | 66532 |
Gene Name: | RAB15 effector protein |
Gene Aliases: |
2210417D09Rik |
Location: |
Chr.6:147032537-147033518 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rep15 | NM_025620.2 | 1 | 325 | NP_079896.1 |
AK008892.1 | 1 | 326 | BAB25956.1 | |
AK008964.1 | 1 | 329 | BAB25994.1 | |
BC024344.1 | 1 | 276 | AAH24344.1 |
