Assay Details
Target Gene Details
Entrez Gene ID: | 20266 |
Gene Name: | sodium channel, voltage-gated, type I, beta |
Gene Aliases: |
- |
Location: |
Chr.7:31116524-31126945 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn1b | NM_011322.2 | 6 | 1415 | NP_035452.1 |
| AK028238.1 | 1 | 654 | BAC25833.1 | |
| BC009652.1 | 6 | 1415 | AAH09652.1 | |
| BC039140.1 | 7 | 1418 | AAH39140.1 | |
| U85786.1 | 6 | 1235 | AAB49368.1 |
More Information
Set Membership: |
|

Genomic Map