Assay Details
Target Gene Details
Entrez Gene ID: | 72431 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 18 |
Gene Aliases: |
2010110O04Rik |
Location: |
Chr.7:43634670-43650228 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam18 | NM_028236.1 | 4 | 745 | NP_082512.1 |
AK008389.1 | 4 | 777 | BAB25642.1 | |
BC042780.1 | 4 | 745 | AAH42780.1 |
