Assay Details
Target Gene Details
Entrez Gene ID: | 319930 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 19 |
Gene Aliases: |
C130022P09Rik, CEAL1 |
Location: |
Chr.7:19875742-19887990 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam19 | NM_177036.5 | 2 | 343 | NP_796010.1 |
AK047942.1 | 2 | 330 | BAC33195.1 | |
AK079290.1 | 2 | 344 | BAC37596.1 | |
AK163794.1 | 2 | 331 | BAE37496.1 | |
BC107227.1 | 2 | 271 | AAI07228.1 | |
BC107228.2 | 2 | 271 | AAI07229.1 | |
BC109333.3 | 2 | 271 | AAI09334.1 |
