Assay Details
Target Gene Details
Entrez Gene ID: | 632778 |
Gene Name: | glutamate rich 4 |
Gene Aliases: |
EG632778, Gm7092, Kseg1 |
Location: |
Chr.7:25614621-25615892 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erich4 | NM_001039243.3 | 2 | 686 | NP_001034332.2 |
AK143993.1 | 2 | 643 | BAE25653.1 | |
BC150835.1 | 2 | 687 |
