Assay Details
Target Gene Details
Entrez Gene ID: | 66245 |
Gene Name: | HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1 |
Gene Aliases: |
1500019G21Rik |
Location: |
Chr.7:4660515-4685121 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hspbp1 | NM_024172.3 | 8 | 1214 | NP_077134.1 |
AF338351.1 | 7 | 1152 | AAK11657.1 | |
AK013263.1 | 8 | 1205 | BAB28756.1 | |
AK088469.1 | 8 | 1190 | BAC40373.1 | |
AK160128.1 | 8 | 1210 | ||
AK209519.1 | 2 | 87 | ||
BC014758.1 | 8 | 1214 | AAH14758.1 |
