Assay Details
Target Gene Details
Entrez Gene ID: | 12406 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade H, member 1 |
Gene Aliases: |
BERF-1, Cbp1, Cbp2, Hsp47, J6, Serpinh2, gp46 |
Location: |
Chr.7:99345375-99353239 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinh1 | NM_001111043.1 | 3 | 454 | NP_001104513.1 |
NM_001111044.1 | 2 | 410 | NP_001104514.1 | |
NM_001285776.1 | 3 | 494 | NP_001272705.1 | |
NM_009825.2 | 2 | 581 | NP_033955.2 | |
AK029080.1 | 3 | 372 | BAC26283.1 | |
AK031457.1 | 2 | 459 | BAC27413.1 | |
AK076474.1 | 2 | 286 | BAC36360.1 | |
AK077660.1 | 2 | 287 | BAC36935.1 | |
AK077937.1 | 2 | 313 | BAC37075.1 | |
AK077992.1 | 3 | 330 | ||
AK079460.1 | 2 | 286 | BAC37652.1 | |
AK086519.1 | 2 | 286 | BAC39683.1 | |
AK086523.1 | 2 | 286 | BAC39684.1 | |
AK089993.1 | 2 | 288 | BAC41032.1 | |
AK090103.1 | 2 | 288 | BAC41093.1 | |
AK159697.1 | 2 | 288 | BAE35297.1 | |
AK159998.1 | 2 | 288 | BAE35546.1 | |
AK166000.1 | 2 | 286 | BAE38510.1 | |
AK167403.1 | 2 | 286 | BAE39492.1 | |
AK168442.1 | 2 | 289 | BAE40347.1 | |
AK168870.1 | 2 | 264 | BAE40689.1 | |
AK169422.1 | 2 | 286 | ||
AK211972.1 | ||||
BC085143.1 | 3 | 327 | AAH85143.1 | |
CA751137.1 | 2 | 459 | ||
J05609.1 | 3 | 328 | AAA03200.1 | |
X60676.1 | 2 | 436 | CAA43091.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
