Assay Details
Target Gene Details
Entrez Gene ID: | 14356 |
Gene Name: | translocase of inner mitochondrial membrane 10B |
Gene Aliases: |
Fxc1, Tim10b, Tim9b |
Location: |
Chr.7:105640250-105641849 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timm10b | NM_019502.2 | 3 | 861 | NP_062375.1 |
| AK003421.1 | 4 | 427 | ||
| AK010994.1 | 4 | 561 | BAB27313.1 | |
| BC009158.1 | 3 | 861 | AAH09158.1 |
More Information
Set Membership: |
|

Genomic Map