Assay Details
Target Gene Details
Entrez Gene ID: | 72040 |
Gene Name: | cadherin-related family member 5 |
Gene Aliases: |
1810074H01Rik, AI481143, Mucdhl, Mupcdh |
Location: |
Chr.7:141269080-141276794 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdhr5 | NM_001114322.1 | 15 | 2280 | NP_001107794.1 |
| NM_028069.3 | 14 | 1794 | NP_082345.1 | |
| AF462391.1 | 15 | 2269 | AAL67856.1 | |
| AK007988.1 | 14 | 1783 | BAB25392.1 | |
| AK027913.1 | 15 | 2281 | BAC25662.1 | |
| BC043336.1 | 15 | 2245 | ||
| BC054469.1 | 14 | 1779 | AAH54469.1 | |
| BG063646.2 | 1 | 350 |
More Information
Set Membership: |
|

Genomic Map