Assay Details
Target Gene Details
Entrez Gene ID: | 57278 |
Gene Name: | basal cell adhesion molecule |
Gene Aliases: |
1200005K12Rik, B-CAM, Gplu, Lu |
Location: |
Chr.7:19756138-19770532 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcam | NM_020486.2 | 15 | 2327 | NP_065232.1 |
AB035511.1 | 15 | 2288 | BAB16053.1 | |
AF221507.1 | 15 | 2277 | AAF34657.1 | |
AK075704.1 | 15 | 2292 | BAC35900.1 | |
AK158439.1 | 15 | 2293 | ||
BB190832.1 | 1 | 110 | ||
BC004826.1 | 15 | 2164 | AAH04826.1 |
