Assay Details
Target Gene Details
Entrez Gene ID: | 232790 |
Gene Name: | osteoclast associated receptor |
Gene Aliases: |
mOSCAR, mOSCAR-M1, mOSCAR-M2, mOSCAR-M3 |
Location: |
Chr.7:3609813-3616157 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oscar | NM_001290377.1 | 4 | 642 | NP_001277306.1 |
| NM_175632.3 | 4 | 660 | NP_783440.1 | |
| AF391159.1 | 4 | 618 | AAL68492.1 | |
| AF391160.1 | 4 | 642 | AAL68493.1 | |
| AF391161.1 | 4 | 636 | AAL68494.1 | |
| BC137776.1 | 4 | 587 |

Genomic Map