Assay Details
Target Gene Details
Entrez Gene ID: | 232790 |
Gene Name: | osteoclast associated receptor |
Gene Aliases: |
mOSCAR, mOSCAR-M1, mOSCAR-M2, mOSCAR-M3 |
Location: |
Chr.7:3609813-3616157 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oscar | NM_001290377.1 | 5 | 1107 | NP_001277306.1 |
NM_175632.3 | 5 | 1125 | NP_783440.1 | |
AF391159.1 | 5 | 1083 | AAL68492.1 | |
AF391161.1 | 5 | 1101 | AAL68494.1 | |
BC137776.1 |
